Please login to your online account to display your discounted pricing

Alfa Aesar™ (Methylthio)acetaldehyde dimethyl acetal, 97%

Chemical Identifiers

Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal
CAS 40015-15-4
Molecular Formula CH3SCH2CH(OCH3)2
Formula Weight 136.21
MDL Number MFCD00009848
UN Number UN1993
Beilstein 773689
Synonym 1,1-Dimethoxy-2-(methylthio)ethane; (Methylmercapto)acetaldehyde dimethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2266706

 
alfa aesar™
B2266706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2266714

 
alfa aesar™
B2266714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.21
MDL Number MFCD00009848
UN Number UN1993
Synonym 1,1-Dimethoxy-2-(methylthio)ethane; (Methylmercapto)acetaldehyde dimethyl acetal
Boiling Point 170° to 171°C
Quantity 5g
Flash Point 53°C (127°F)
Health Hazard 2 GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal
Assay 97%
Molecular Formula CH3SCH2CH(OCH3)2
CAS 40015-15-4
Beilstein 773689
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.