missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylsulfoxide, 99.9% min., OmniSolv, EMD Millipore

Manufacturer:  Millipore Sigma MX14566

 View more versions of this product

Catalog No. MMX14566



Specifications

Specifications

Methylsulfoxide
214.85°C (418.7°F)
0.053hPa (20°C)
Colorless
1L
99.9% min.
(CH3)2SO
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.1
1.10g/cm3 at 20°C
Glass Bottle
189°C
18.4°C
67-68-5
C2H6OS
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(=O)C
78.129
CHEBI:28262
Liquid

Chemical Identifiers

67-68-5
78.129
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(=O)C
C2H6OS
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to +25°C

Documents