missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylsulfoxide, 99.8% min., DriSolv™, EMD Millipore

$103.49 - $226.46

Chemical Identifiers

CAS 67-68-5
Physical Form Liquid
Assay Percent Range 99.8%, min.
Molecular Formula (CH3)2SO
Linear Formula (CH3)2SO
Formula Weight 78.14
Synonym Dimethyl Sulfoxide
Chemical Name or Material Methylsulfoxide, DriSolv™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MMX14576 Emd Millipore Corporation
MX14576
1L Each for $226.46
MMX14577 Emd Millipore Corporation
MX14577
100mL Each for $103.49
Specifications Description & Specifications

Chemical Identifiers

67-68-5
99.8%, min.
(CH3)2SO
Dimethyl Sulfoxide
Liquid
(CH3)2SO
78.14
Methylsulfoxide, DriSolv™

Specifications

78.14
Clear
(CH3)2SO
99.8%, min.
Dimethyl Sulfoxide
1L
Liquid
67-68-5
(CH3)2SO
Methylsulfoxide, DriSolv™

Safety and Handling

Recommended Storage : Room Temperature