missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylsulfonylacetonitrile, TCI America™

$67.22 - $192.50

Chemical Identifiers

CAS 2274-42-2
Molecular Formula C3H5NO2S
Molecular Weight (g/mol) 119.138
MDL Number MFCD00014743
InChI Key FOTRKCAZUSJCQD-UHFFFAOYSA-N
Synonym methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
PubChem CID 75283
ChEBI CHEBI:32444
IUPAC Name 2-methylsulfonylacetonitrile
SMILES CS(=O)(=O)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M06675G View Documents TCI America
M06675G
5g Each for $67.22
 
M066725G View Documents TCI America
M066725G
25g Each for $192.50
 
Specifications

Chemical Identifiers

2274-42-2
119.138
FOTRKCAZUSJCQD-UHFFFAOYSA-N
75283
2-methylsulfonylacetonitrile
C3H5NO2S
MFCD00014743
methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
CHEBI:32444
CS(=O)(=O)CC#N

Specifications

White-Yellow
5g
C3H5NO2S
3439
FOTRKCAZUSJCQD-UHFFFAOYSA-N
2-methylsulfonylacetonitrile
75283
119.14
≥98.0% (GC)
84°C
2274-42-2
MFCD00014743
methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
CS(=O)(=O)CC#N
119.138
CHEBI:32444
Crystal-Powder at 20°C
Methylsulfonylacetonitrile
Safety and Handling

Safety and Handling

TSCA : No