Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylsulfonylacetonitrile, 98+%

$173.28 - $679.55

Chemical Identifiers

Chemical Name or Material Methylsulfonylacetonitrile
CAS 2274-42-2
Molecular Formula C3H5NO2S
Formula Weight 119.14
MDL Number MFCD00014743
Beilstein 969929
Synonym Methanesulfonylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0328009

  SDS
alfa aesar™
L0328009
10g Each for $173.28

AAL0328018

  SDS
alfa aesar™
L0328018
50g Each for $679.55
Description & Specifications

Specifications

Formula Weight 119.14
Molecular Formula C3H5NO2S
MDL Number MFCD00014743
CAS 2274-42-2
Synonym Methanesulfonylacetonitrile
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Chemical Name or Material Methylsulfonylacetonitrile
Assay ≥98%
Beilstein 969929
Health Hazard 1 Warning
Melting Point 82° to 86°C
Quantity 10g
Health Hazard 3 GHS P Statement
P280h-P302+P352
Wear protective gloves/clothing.
IF ON SKIN: Wash with plenty of soap and water.

Applications
(Methylsulfonyl)acetonitrile was used in the synthesis of substituted δ-pyrone ring analogues, 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P280h-P302+P352
Wear protective gloves/clothing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.