missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylsulfonylacetone, 98+%

$38.42 - $432.34

Chemical Identifiers

CAS 5000-46-4
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.165
MDL Number MFCD00014745
InChI Key NWEYGXQKFVGUFR-UHFFFAOYSA-N
Synonym 1-methanesulfonylpropan-2-one, 1-methylsulfonyl acetone, 1-methylsulfonyl propan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methanesulfonylacetone, methanesulphonylacetone, methylsulfonylaceton, methylsulfonylacetone, methylsulphonylacetone
PubChem CID 78695
IUPAC Name 1-methylsulfonylpropan-2-one
SMILES CC(=O)CS(=O)(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0342104
View Documents
Alfa Aesar™
L0342104
2g Each for $38.42
 
AAL0342109
View Documents
Alfa Aesar™
L0342109
10g Each for $115.57
 
AAL0342118
View Documents
Alfa Aesar™
L0342118
50g Each for $432.34
 
Description

Description

Applications
Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents
Specifications

Chemical Identifiers

5000-46-4
136.165
NWEYGXQKFVGUFR-UHFFFAOYSA-N
78695
CC(=O)CS(=O)(=O)C
C4H8O3S
MFCD00014745
1-methanesulfonylpropan-2-one, 1-methylsulfonyl acetone, 1-methylsulfonyl propan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methanesulfonylacetone, methanesulphonylacetone, methylsulfonylaceton, methylsulfonylacetone, methylsulphonylacetone
1-methylsulfonylpropan-2-one

Specifications

47°C to 52°C
5000-46-4
MFCD00014745
1-methanesulfonylpropan-2-one, 1-methylsulfonyl acetone, 1-methylsulfonyl propan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methanesulfonylacetone, methanesulphonylacetone, methylsulfonylaceton, methylsulfonylacetone, methylsulphonylacetone
NWEYGXQKFVGUFR-UHFFFAOYSA-N
1-methylsulfonylpropan-2-one
78695
≥98%
2g
C4H8O3S
1754113
Insoluble in water.
CC(=O)CS(=O)(=O)C
136.165
136.18
Methylsulfonylacetone
Safety and Handling

Safety and Handling

EINECSNumber : 225-665-6

TSCA : No

Recommended Storage : Ambient temperatures