Learn More
Methylsulfonylacetone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0342109
Description
Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents
Specifications
Methylsulfonylacetone | |
47°C to 52°C | |
MFCD00014745 | |
1754113 | |
Insoluble in water. | |
CC(=O)CS(=O)(=O)C | |
136.165 | |
136.18 |
5000-46-4 | |
C4H8O3S | |
10 g | |
methylsulfonylacetone, 1-methylsulfonyl propan-2-one, methanesulfonylacetone, 1-methylsulfonyl acetone, 1-methanesulfonylpropan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methylsulfonylaceton, methylsulphonylacetone, methanesulphonylacetone | |
NWEYGXQKFVGUFR-UHFFFAOYSA-N | |
1-methylsulfonylpropan-2-one | |
78695 | |
≥98% |
Chemical Identifiers
5000-46-4 | |
136.165 | |
NWEYGXQKFVGUFR-UHFFFAOYSA-N | |
78695 | |
CC(=O)CS(=O)(=O)C |
C4H8O3S | |
MFCD00014745 | |
methylsulfonylacetone, 1-methylsulfonyl propan-2-one, methanesulfonylacetone, 1-methylsulfonyl acetone, 1-methanesulfonylpropan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methylsulfonylaceton, methylsulphonylacetone, methanesulphonylacetone | |
1-methylsulfonylpropan-2-one |
Safety and Handling
EINECSNumber : 225-665-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only