missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylsulfonylacetone, 98+%

Manufacturer:  Alfa Aesar™ L0342104

 View more versions of this product

Catalog No. AAL0342104



Description

Description

Applications
Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents
Specifications

Specifications

Methylsulfonylacetone
2g
C4H8O3S
1754113
NWEYGXQKFVGUFR-UHFFFAOYSA-N
1-methylsulfonylpropan-2-one
78695
≥98%
47°C to 52°C
5000-46-4
MFCD00014745
methylsulfonylacetone, 1-methylsulfonyl propan-2-one, methanesulfonylacetone, 1-methylsulfonyl acetone, 1-methanesulfonylpropan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methylsulfonylaceton, methylsulphonylacetone, methanesulphonylacetone
CC(=O)CS(=O)(=O)C
136.165
136.18

Chemical Identifiers

5000-46-4
136.165
NWEYGXQKFVGUFR-UHFFFAOYSA-N
78695
CC(=O)CS(=O)(=O)C
C4H8O3S
MFCD00014745
methylsulfonylacetone, 1-methylsulfonyl propan-2-one, methanesulfonylacetone, 1-methylsulfonyl acetone, 1-methanesulfonylpropan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methylsulfonylaceton, methylsulphonylacetone, methanesulphonylacetone
1-methylsulfonylpropan-2-one
Documents