Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylsulfonylacetic acid, 98+%

$215.47 - $215.47

Chemical Identifiers

Chemical Name or Material Methylsulfonylacetic acid
CAS 2516-97-4
Molecular Formula C3H6O4S
Formula Weight 138.14
MDL Number MFCD00051843
Beilstein 1760342
Synonym Methanesulfonylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0759806

  SDS
alfa aesar™
L0759806
5g Each for $215.47
Description & Specifications

Specifications

Formula Weight 138.14
MDL Number MFCD00051843
Assay ≥98%
Synonym Methanesulfonylacetic acid
Molecular Formula C3H6O4S
Melting Point 114° to 119°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2516-97-4
Beilstein 1760342
Chemical Name or Material Methylsulfonylacetic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 5g

Applications
Methylsulfonylacetic acid is used as a pharmaceutical intermediate .

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.