missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylsulfamic Acid, TCI America™

$227.64 - $902.94

Chemical Identifiers

CAS 4112-03-2
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.12
MDL Number MFCD00010870
InChI Key MYMDOKBFMTVEGE-UHFFFAOYSA-N
PubChem CID 244258
IUPAC Name methylsulfamic acid
SMILES CNS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10835G View Documents TCI America
M10835G
5g Each for $227.64
 
M108325G View Documents TCI America
M108325G
25g Each for $902.94
 
Specifications

Chemical Identifiers

4112-03-2
111.12
MYMDOKBFMTVEGE-UHFFFAOYSA-N
methylsulfamic acid
CH5NO3S
MFCD00010870
244258
CNS(=O)(=O)O

Specifications

White-Yellow
4112-03-2
MFCD00010870
MYMDOKBFMTVEGE-UHFFFAOYSA-N
methylsulfamic acid
244258
Crystal-Powder at 20°C
Methylsulfamic Acid
5g
CH5NO3S
1759
CNS(=O)(=O)O
111.12
111.12
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No