Please login to your online account to display your discounted pricing

Methylsulfamic acid, 98%, ACROS Organics™

$92.16 - $92.16

Chemical Identifiers

Chemical Name or Material Methylsulfamic acid
CAS 807979
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula CH5NO3S
Linear Formula CH3NHSO3H
Formula Weight 111.12
MDL Number MFCD00010870
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314670250

  SDS
thermo fisher scientific
AC314670250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC314670050

  SDS
thermo fisher scientific
314670050
Glass bottle 5g Each for $92.16
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Assay ≥97.5%
Formula Weight 111.12
Quantity 25g
Color White
Melting Point 189° to 193°C
CAS 807979
CAS Max % 100.0
Chemical Name or Material Methylsulfamic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula CH5NO3S
Linear Formula CH3NHSO3H
Assay Percent Range 98%
MDL Number MFCD00010870
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.