missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylsulfamic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 314670050

 View more versions of this product

Catalog No. AC314670050


 

Specifications Description & Specifications

Specifications

≥97.5%
111.12
White
807979
Methylsulfamic acid
98%
MFCD00010870
Authentic
Crystals or Crystalline Powder
5g
189° to 193°C
100.0
CH5NO3S
97.5
Glass bottle
CH3NHSO3H

Chemical Identifiers

807979
100.0
CH5NO3S
111.12
Methylsulfamic acid
97.5
98%
CH3NHSO3H
MFCD00010870
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger