missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylsuccinic acid, 99%

Manufacturer:  Alfa Aesar™ H6096714

 View more versions of this product

Catalog No. AAH6096714



Description

Description

Applications
Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
Specifications

Specifications

Methylsuccinic acid
25g
C5H8O4
MFCD00002659
1,2-propanedicarboxylic acid, 2-methyl-butanedioic acid, 2-methylsuccinic acid, butanedioic acid, 2-methyl, butanedioic acid, methyl, ccris 6068, methyl succinic acid, methylsuccinic acid, pyrotartaric acid, succinic acid, methyl
WXUAQHNMJWJLTG-UHFFFAOYSA-N
2-methylbutanedioic acid
10349
99%
110°C to 115°C
498-21-5
HO2CCH2CH(CH3)CO2H
1722946
Soluble in water.
CC(CC(=O)O)C(=O)O
132.115
132.11

Chemical Identifiers

498-21-5
132.115
WXUAQHNMJWJLTG-UHFFFAOYSA-N
10349
CC(CC(=O)O)C(=O)O
C5H8O4
MFCD00002659
1,2-propanedicarboxylic acid, 2-methyl-butanedioic acid, 2-methylsuccinic acid, butanedioic acid, 2-methyl, butanedioic acid, methyl, ccris 6068, methyl succinic acid, methylsuccinic acid, pyrotartaric acid, succinic acid, methyl
2-methylbutanedioic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 207-857-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents