Please login to your online account to display your discounted pricing

Methylphenylvinylsilane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methylphenylvinylsilane
CAS 17878-39-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H12Si
Linear Formula C6H5Si(H)(CH3)CHCH
Formula Weight 148.28
MDL Number MFCD00053689
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336680050

  SDS
thermo fisher scientific
336680050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 148.28
Quantity 5g
Color Clear colorless
EINECS Number 241-830-5
TSCA TSCA
Density 0.89g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Methylphenylvinylsilane
Refractive Index 1.5125 to 1.5145 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C9H12Si
Linear Formula C6H5Si(H)(CH3)CHCH
MDL Number MFCD00053689
CAS 17878-39-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Reduction reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.