Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylphenylphosphinic acid, 98%

$45.81 - $629.82

Chemical Identifiers

Chemical Name or Material Methylphenylphosphinic acid
CAS 4271-13-0
Molecular Formula C7H9O2P
Formula Weight 156.12
MDL Number MFCD00123313
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1977403

  SDS
alfa aesar™
A1977403
1g Each for $45.81

AAA1977406

  SDS
alfa aesar™
A1977406
5g Each for $162.73

AAA1977414

  SDS
alfa aesar™
A1977414
25g Each for $629.82
Description & Specifications

Specifications

Formula Weight 156.12
Chemical Name or Material Methylphenylphosphinic acid
Assay 98%
Molecular Formula C7H9O2P
MDL Number MFCD00123313
CAS 4271-13-0
Health Hazard 1 Warning
Melting Point 132° to 134°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.