Please login to your online account to display your discounted pricing

β-Methylphenethylamine, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  177240250

 View more versions of this product

Catalog No. AC177240250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 135.21
Quantity 25g
Color Clear colorless
EINECS Number 209-479-2
TSCA TSCA
RTECS Number SH9520000
Density 0.9300g/mL
CAS Max % 100.0
Refractive Index 1.5230 to 1.5250 (20°C, 589nm)
Synonym 1-Amino-2-phenylpropane
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material β-Methylphenethylamine
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H13N
Linear Formula C6H5CH(CH3)CH2NH2
MDL Number MFCD00008144
CAS 582-22-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material β-Methylphenethylamine
Name Note 99%
CAS 582-22-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H13N
Linear Formula C6H5CH(CH3)CH2NH2
Formula Weight 135.21
MDL Number MFCD00008144
Synonym 1-Amino-2-phenylpropane
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents