Please login to your online account to display your discounted pricing

β-Methylphenethylamine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material β-Methylphenethylamine
Name Note 99%
CAS 582-22-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H13N
Linear Formula C6H5CH(CH3)CH2NH2
Formula Weight 135.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177240250

  SDS
thermo fisher scientific
177240250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 135.21
Quantity 25g
Color Clear colorless
TSCA TSCA
RTECS Number SH9520000
EINECS Number 209-479-2
Density 0.9300g/mL
CAS Max % 100.0
Packaging Glass bottle
Synonym 1-Amino-2-phenylpropane
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS Min % 98.5
Refractive Index 1.5230 to 1.5250 (20°C, 589nm)
Name Note 99%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Chemical Name or Material β-Methylphenethylamine
Molecular Formula C9H13N
MDL Number MFCD00008144
Linear Formula C6H5CH(CH3)CH2NH2
CAS 582-22-9
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.