Please login to your online account to display your discounted pricing

Methylmercury(II) chloride, standard solution in H2O, Alfa Aesar™

$203.42 - $399.30

Chemical Identifiers

Chemical Name or Material Methylmercury(II) chloride
CAS 115-09-3
Physical Form Liquid
Molecular Formula CH3HgCl
Formula Weight 251.08
MDL Number MFCD00013592
Synonym Chloromethylmercury
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA33553AA

  SDS
alfa aesar
33553AA
5mL Each for $203.42

AA33553AC

  SDS
alfa aesar
33553AC
25mL Each for $399.30
Description & Specifications

Specifications

Formula Weight 251.08
Quantity 5mL
Color Clear
DOT Information DOT Class: 6.1; Packing Group: II
EINECS Number 204-064-2
Molecular Formula CH3HgCl
Physical Form Liquid
Appearance Clear
CAS 115-09-3
MDL Number MFCD00013592
Synonym Chloromethylmercury
Chemical Name or Material Methylmercury(II) chloride
Health Hazard 1 H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Safety and Handling

H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.