missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylmalonic Acid, TCI America™

Manufacturer:  TCI America M056810G

 View more versions of this product

Catalog No. M056810G



Specifications

Specifications

Methylmalonic Acid
135°C
516-05-2
MFCD00002656
ZIYVHBGGAOATLY-UHFFFAOYSA-N
2-methylpropanedioic acid
487
118.09
≥98.0% (T)
White
10g
C4H6O4
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
CC(C(=O)O)C(=O)O
118.088
CHEBI:30860
Crystal-Powder at 20°C

Chemical Identifiers

516-05-2
118.088
ZIYVHBGGAOATLY-UHFFFAOYSA-N
487
2-methylpropanedioic acid
C4H6O4
MFCD00002656
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
CHEBI:30860
CC(C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : OO1400000

TSCA : No

Documents