missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylmalonic acid, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 127090250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methylmalonic acid | |
95.0 | |
1.4500g/mL | |
96% | |
C4H6O4 | |
MFCD00002656 | |
02, IV, 1932 | |
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7 | |
ZIYVHBGGAOATLY-UHFFFAOYSA-N | |
2-methylpropanedioic acid | |
487 | |
118.09 |
516-05-2 | |
100.0 | |
Authentic | |
Glass bottle | |
HO2CCH(CH3)CO2H | |
25g | |
1.45 | |
Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid | |
CC(C(O)=O)C(O)=O | |
118.09 | |
CHEBI:30860 | |
96% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 208-219-5
RUO â Research Use Only