Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material Methylmagnesium chloride
CAS 676-58-4
Linear Formula CH3MgCl
Formula Weight 74.81
MDL Number MFCD00000468
UN Number UN3399
Beilstein 3587199
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42101JS

  SDS
alfa aesar™
42101JS
0.25 mole CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.81
Chemical Name or Material Methylmagnesium chloride
CAS 676-58-4
UN Number UN3399
Beilstein 3587199
Health Hazard 1 Danger
Concentration 3M in THF
Quantity 0.25 mole
Sensitivity Air & Moisture Sensitive
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Density 1.013g/mL
TSCA TSCA
Packaging ChemSeal™ bottles
Physical Form Liquid
Linear Formula CH3MgCl
MDL Number MFCD00000468
Color Pale brown to dark green
Flash Point −17° (1°F)
Odor Ether-like
Health Hazard 2 GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Packaged under argon