Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylmagnesium chloride, 3M in THF

As a grignard reagent and an alkylating agent in organic synthesis

Chemical Identifiers

Chemical Name or Material Methylmagnesium chloride
CAS 676-58-4
Molecular Formula CH3ClMg
Linear Formula CH3MgCl
Formula Weight 74.81
MDL Number MFCD00000468
UN Number UN3399
Beilstein 3587199
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA46118JS

  SDS
alfa aesar™
46118JS
0.25 mole CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.81
Sensitivity Air & Moisture Sensitive
Linear Formula CH3MgCl
Color Pale brown to dark green
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Beilstein 3587199
Solubility Information Insoluble in hydrocarbons
MDL Number MFCD00000468
Health Hazard 1 Danger
Molecular Formula CH3ClMg
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
TSCA TSCA
Concentration 3M in THF
CAS 676-58-4
UN Number UN3399
Chemical Name or Material Methylmagnesium chloride
Quantity 0.25 mole
Odor Ether-like
Health Hazard 2 GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Physical Form Liquid
Density 1.013g/mL
Flash Point −17°C (1°F)
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.