missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylmagnesium chloride, 3M in THF

As a grignard reagent and an alkylating agent in organic synthesis

Manufacturer:  Alfa Aesar™ 46118JS

 View more versions of this product

Catalog No. AA46118JS



Specifications

Specifications

Methylmagnesium chloride
−17°C (1°F)
Air & Moisture Sensitive
0.25 mol
CH3ClMg
MFCD00000468
3587199
3M in THF
CCERQOYLJJULMD-UHFFFAOYSA-M
magnesium;carbanide;chloride
12670
Liquid
1.013g/mL
Ether-like
Brown to Green
676-58-4
CH3MgCl
UN3399
methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
Insoluble in hydrocarbons
[CH3-].[Mg+2].[Cl-]
74.79
74.81
3M in THF

Chemical Identifiers

676-58-4
74.79
CCERQOYLJJULMD-UHFFFAOYSA-M
12670
[CH3-].[Mg+2].[Cl-]
CH3ClMg
MFCD00000468
methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
magnesium;carbanide;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

SDS Documents