Learn More
Methylmagnesium bromide, 1M solution in CPME, AcroSeal™, Thermo Scientific™
Supplier: Thermo Fisher Scientific 446008000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methylmagnesium bromide | |
800mL | |
CH3MgBr | |
−1°C | |
Solubility in water: reacts | |
[CH3-].[Mg++].[Br-] | |
119.24 | |
119.24 |
75-16-1,5614-37-9 | |
CH3BrMg | |
MFCD00000041 | |
bromo methyl magnesium, ch3mgbr, grignard reagent, magnesium, bromomethyl, memgbr, methyl magnesium bromide, methylmagnesium bromide, methylmagnesium bromide solution, 3.0 m in diethyl ether, methylmagnesiumbromide, methymagnesiumbromide, unii-22cw9773df | |
NXPHGHWWQRMDIA-UHFFFAOYSA-M | |
magnesium;carbanide;bromide | |
6349 |
Chemical Identifiers
75-16-1 | |
119.24 | |
NXPHGHWWQRMDIA-UHFFFAOYSA-M | |
6349 | |
[CH3-].[Mg++].[Br-] |
CH3BrMg | |
MFCD00000041 | |
bromo methyl magnesium, ch3mgbr, grignard reagent, magnesium, bromomethyl, memgbr, methyl magnesium bromide, methylmagnesium bromide, methylmagnesium bromide solution, 3.0 m in diethyl ether, methylmagnesiumbromide, methymagnesiumbromide, unii-22cw9773df | |
magnesium;carbanide;bromide |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 200-844-1
RUO – Research Use Only