Learn More
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 183541000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methylmagnesium bromide | |
3M Solution in Diethyl Ether | |
CH3BrMg | |
MFCD00000041 | |
1.0350g/mL | |
Solubility in water: vigorous reaction. | |
C[Mg]Br | |
119.24 | |
119.26 | |
1.035 | |
−40°C |
Colorless to Yellow | |
75-16-1,60-29-7 | |
CH3MgBr | |
2.85 to 3.20M | |
methylmagnesium bromide, grignard reagent, methyl magnesium bromide, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, methylmagnesium bromide solution, 3.0 m in diethyl ether, memgbr, memgbr, methymagnesiumbromide, methylmagnesiumbromide, methylmagnesiumbromide | |
AVFUHBJCUUTGCD-UHFFFAOYSA-M | |
magnesium;carbanide;bromide | |
6349 | |
AcroSeal™ Glass bottle | |
100 mL | |
Solution, may develop some turbidity precipitate |
Chemical Identifiers
75-16-1 | |
119.24 | |
AVFUHBJCUUTGCD-UHFFFAOYSA-M | |
6349 |
CH3BrMg | |
MFCD00000041 | |
methylmagnesium bromide, grignard reagent, methyl magnesium bromide, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, methylmagnesium bromide solution, 3.0 m in diethyl ether, memgbr, memgbr, methymagnesiumbromide, methylmagnesiumbromide, methylmagnesiumbromide | |
C[Mg]Br |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Danger
RUO – Research Use Only