Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylmagnesium bromide, 3M in ether

$204.92 - $204.92

Chemical Identifiers

Chemical Name or Material Methylmagnesium bromide
CAS 75-16-1
Linear Formula CH3MgBr
Formula Weight 119.26
MDL Number MFCD00000041
UN Number UN1928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA87324AP

  SDS
alfa aesar™
87324AP
500mL Each for $204.92
Description & Specifications

Specifications

Chemical Name or Material Methylmagnesium bromide
CAS 75-16-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Density 1.035g/mL
DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: METHYL MAGNESIUM BROMIDE, IN ETHYL ETHER
TSCA TSCA
Physical Form Liquid
Linear Formula CH3MgBr
Formula Weight 119.26
MDL Number MFCD00000041
UN Number UN1928
Color Colorless to pale brown
Quantity 500mL
Odor Ether-like
Concentration 3M in ether
Sensitivity Air & Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.