missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylmagnesium bromide, 3M in ether

Manufacturer:  Alfa Aesar™ 87324AP

 View more versions of this product

Catalog No. AA87324AP



Specifications

Specifications

Methylmagnesium bromide
Ether-like
Brown to Colorless
75-16-1
CH3MgBr
UN1928
3M in ether
[CH3-].[Mg+2].[Br-]
119.244
119.26
3M in ether
1.035g/mL
Air & Moisture Sensitive
500mL
CH3BrMg
MFCD00000041
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
NXPHGHWWQRMDIA-UHFFFAOYSA-M
magnesium;carbanide;bromide
6349
Liquid

Chemical Identifiers

75-16-1
119.244
NXPHGHWWQRMDIA-UHFFFAOYSA-M
6349
[CH3-].[Mg+2].[Br-]
CH3BrMg
MFCD00000041
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
magnesium;carbanide;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: METHYL MAGNESIUM BROMIDE, IN ETHYL ETHER

TSCA : TSCA

SDS Documents