Please login to your online account to display your discounted pricing

Methylmagnesium bromide, 1.4M solution in Toluene/THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC377338000

 View more versions of this product

Catalog No. AC377338000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of damaging the unborn child. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. Suspected of causing cancer. May form explosive peroxides.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Physical Form Solution
Formula Weight 119.24
Quantity 800mL
Color Light yellow or tan to dark brown
Packaging AcroSeal™ Glass Bottle
Concentration 1.20 to 1.55M
Density 1.0200g/mL
Linear Formula CH3MgBr
CAS 109-99-9
MDL Number MFCD00000041
CAS 75-16-1
Chemical Name or Material Methylmagnesium bromide
Additional Information product may develop some turbidity or precipitate
CAS 108-88-3
Health Hazard 1 Danger
Name Note 1.4M Solution in Toluene/THF
Molecular Formula CH3BrMg

Specifications

Chemical Name or Material Methylmagnesium bromide
Name Note 1.4M Solution in Toluene/THF
CAS 75-16-1
CAS 109-99-9
CAS 108-88-3
Molecular Formula CH3BrMg
Linear Formula CH3MgBr
Formula Weight 119.24
MDL Number MFCD00000041
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of damaging the unborn child. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. Suspected of causing cancer. May form explosive peroxides.

SDS & Safety Documents