Please login to your online account to display your discounted pricing

Methylmagnesium bromide, 1.4M solution in Toluene/THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC377331000

 View more versions of this product

Catalog No. AC377331000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 119.24
Quantity 100mL
Color Light yellow or tan to dark brown
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 1.20 to 1.55M
Density 1.0200g/mL
Linear Formula CH3MgBr
Health Hazard 2 GHS P Statement May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of
Health Hazard 3 GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou
Chemical Name or Material Methylmagnesium bromide
Name Note 1.4M Solution in Toluene/THF
Molecular Formula CH3BrMg
CAS 75-16-1
Additional Information product may develop some turbidity or precipitate
CAS 109-99-9
MDL Number MFCD00000041
Health Hazard 1 Danger
CAS 108-88-3

Specifications

Chemical Name or Material Methylmagnesium bromide
Name Note 1.4M Solution in Toluene/THF
CAS 75-16-1
CAS 109-99-9
CAS 108-88-3
Molecular Formula CH3BrMg
Linear Formula CH3MgBr
Formula Weight 119.24
MDL Number MFCD00000041
Accessories

Danger

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

GHS P Statement May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of

SDS & Safety Documents