missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylmagnesium bromide, 1.4M solution in Toluene/THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.244
MDL Number MFCD00000041
InChI Key NXPHGHWWQRMDIA-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
PubChem CID 6349
IUPAC Name magnesium;carbanide;bromide
SMILES [CH3-].[Mg+2].[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377331000 SDS View Documents Acros Organics
AC377331000
100mL AcroSeal™ Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377338000 SDS View Documents Acros Organics
AC377338000
800mL AcroSeal™ Glass Bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

75-16-1
119.244
NXPHGHWWQRMDIA-UHFFFAOYSA-M
6349
[CH3-].[Mg+2].[Br-]
CH3BrMg
MFCD00000041
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
magnesium;carbanide;bromide

Specifications

75-16-1,109-99-9,108-88-3
CH3BrMg
MFCD00000041
1.0200g/mL
NXPHGHWWQRMDIA-UHFFFAOYSA-M
magnesium;carbanide;bromide
6349
Solution
Solubility in water: vigorous reaction.
Brown to Tan or Yellow
Methylmagnesium bromide, 1.4M Solution in Toluene/THF
product may develop some turbidity or precipitate
CH3MgBr
1.20 to 1.55M
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
[CH3-].[Mg+2].[Br-]
119.244
119.24
AcroSeal™ Glass bottle
1.02
100mL
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of damaging the unborn child. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water. Suspected of causing cancer. May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger