Please login to your online account to display your discounted pricing

Methyllycaconitine citrate, Tocris

Alpha7 neuronal nicotinic receptor antagonist

$113.57 - $478.51

Chemical Identifiers

Physical Form Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

102925

 
r & d systems
102925
25mg Each for $478.51

10295R

 
r & d systems
10295
5mg Each for $113.57
Description & Specifications

Specifications

Chemical Name or Material Methyllycaconitine citrate
CAS 112825-05-5
Recommended Storage Desiccate at -20°C
Physical Form Solid
Assay Percent Range >98%
Molecular Formula C37H50N2O10·C6H8O7
Formula Weight 874.93
Synonym [1α,4(S),6β,14α,16β]-20-Ethyl-1,6,14,16-tetramethoxy-4-[[[2-(3-methyl-2,5-dioxo-1-pyrrolidinyl)benzoyl]oxy]methyl]aconitane-7,8-diol citrate
Quantity 25mg
Safety and Handling

Desiccate at -20°C