Please login to your online account to display your discounted pricing

Methyllithium lithium bromide complex, 2.2M sol. in diethylether, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyllithium lithium bromide complex
CAS 1309980-11-7
CAS 60-29-7
Assay 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Molecular Formula CH3Li·LiBr
Linear Formula CH3Li·LiBr
Formula Weight 108.82
MDL Number MFCD00044645
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC181291000

  SDS
thermo fisher scientific
AC181291000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC181298000

  SDS
thermo fisher scientific
AC181298000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 108.82
Quantity 100mL
CAS 917-54-4
Color Colorless to yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
Assay 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Density 0.81g/mL
Linear Formula CH3Li·LiBr
Health Hazard 1 GHS Signal Word: Danger
CAS 60-29-7
Chemical Name or Material Methyllithium lithium bromide complex
Storage Information Product may develop some turbidity or precipitate
Molecular Formula CH3Li·LiBr
MDL Number MFCD00044645
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Extremely flammable liquid and v
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Extremely flammable liquid and v