missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyllithium, 3% solution in 2-MeTHF/cumene, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445848000

 View more versions of this product

Catalog No. AC445848000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyllithium
CH3Li
lithium carbanide, lithium methanide, lithium methide, lithium methyl, lithium, methyl, meli, methllithium, methyl lithium, methyllithium, methyllithum
[Li+].[CH3-]
21.96
21.96
2.7 to 3.4% (as MeLi) (Total base)
AcroSeal™ Glass bottle
Colorless
−11°C
917-54-4,98-82-8,96-47-9
CH3Li
IHLVCKWPAMTVTG-UHFFFAOYSA-N
lithium;carbanide
2724049
Liquid
0.8670g/mL
0.867
800mL

Chemical Identifiers

917-54-4
21.96
lithium carbanide, lithium methanide, lithium methide, lithium methyl, lithium, methyl, meli, methllithium, methyl lithium, methyllithium, methyllithum
lithium;carbanide
CH3Li
IHLVCKWPAMTVTG-UHFFFAOYSA-N
2724049
[Li+].[CH3-]
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory i

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place.
Store in a closed container.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Danger

EINECSNumber : 213-026-4

SDS
Documents