missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyllithium, 3% solution in 2-MeTHF/cumene, AcroSeal™, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445841000
Specifications
Methyllithium | |
917-54-4,98-82-8,96-47-9 | |
CH3Li | |
IHLVCKWPAMTVTG-UHFFFAOYSA-N | |
lithium;carbanide | |
2724049 | |
Liquid | |
0.8670g/mL | |
Colorless | |
−11°C |
0.867 | |
CH3Li | |
lithium carbanide, lithium methanide, lithium methide, lithium methyl, lithium, methyl, meli, methllithium, methyl lithium, methyllithium, methyllithum | |
[Li+].[CH3-] | |
21.96 | |
21.96 | |
2.7 to 3.4% (as MeLi) (Total base) | |
AcroSeal™ Glass bottle | |
100mL |
Chemical Identifiers
917-54-4 | |
21.96 | |
lithium carbanide, lithium methanide, lithium methide, lithium methyl, lithium, methyl, meli, methllithium, methyl lithium, methyllithium, methyllithum | |
lithium;carbanide |
CH3Li | |
IHLVCKWPAMTVTG-UHFFFAOYSA-N | |
2724049 | |
[Li+].[CH3-] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory i
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place.
Store in a closed container.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do
GHS Signal Word: Danger
EINECSNumber : 213-026-4