Please login to your online account to display your discounted pricing

Methyllithium, 3% solution in 2-MeTHF/cumene, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyllithium
CAS 917-54-4
CAS 98-82-8
CAS 96-47-9
Assay 2.7 to 3.4% (as MeLi) (Total base)
Molecular Formula CH3Li
Linear Formula CH3Li
Formula Weight 21.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC445848000

 
thermo fisher scientific
AC445848000
Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445841000

 
thermo fisher scientific
AC445841000
Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 2.7 to 3.4% (as MeLi) (Total base)
Physical Form Liquid
Formula Weight 21.96
Quantity 800mL
Color Clear colorless
Packaging Glass Bottle
Density 0.8670g/mL
Health Hazard 1 GHS Signal Word: Danger
CAS 98-82-8
CAS 96-47-9
Molecular Formula CH3Li
Chemical Name or Material Methyllithium
CAS 917-54-4
Linear Formula CH3Li
Safety and Handling

GHS Signal Word: Danger