Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyllithium, 1-2M in ether

Used in Grignard-type reactions

Chemical Identifiers

Chemical Name or Material Methyllithium
CAS 917-54-4
Formula Weight 21.97
MDL Number MFCD00008253
UN Number UN3394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA89693AE

  SDS
alfa aesar™
89693AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 21.97
Chemical Name or Material Methyllithium
Odor Ether-like
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
TSCA TSCA
Concentration 1 to 2M in ether
Health Hazard 2 GHS H Statement
H224-H250-H260-H252-H314-H318-H302-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Quantity 100mL
Sensitivity Air & Moisture Sensitive
MDL Number MFCD00008253
Physical Form Liquid
DOT Information Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Health Hazard 1 Danger
Color Colorless
CAS 917-54-4
UN Number UN3394
Density 0.732g/mL
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H224-H250-H260-H252-H314-H318-H302-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.