missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylhydrazine Sulfate 98.0+%, TCI America™

Supplier:  TCI America M034125G

 View more versions of this product

Catalog No. M034125G


Description

Description

Specifications

Specifications

Methylhydrazine Sulfate
143°C
CH8N2O4S
25 g
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate
CN[NH3+].OS([O-])(=O)=O
144.15
144.15
Crystalline Powder
302-15-8
White-Yellow
MFCD00035423
2811
KJDJPXUIZYHXEZ-UHFFFAOYSA-N
2-methylhydrazinium hydrogen sulfate
165286
≥98.0% (T)

Chemical Identifiers

302-15-8
144.15
KJDJPXUIZYHXEZ-UHFFFAOYSA-N
165286
CN[NH3+].OS([O-])(=O)=O
CH8N2O4S
MFCD00035423
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate
2-methylhydrazinium hydrogen sulfate
Safety and Handling

Safety and Handling

RTECSNumber : MV7750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions