Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylhydrazine sulfate, 98%

Chemical Identifiers

Chemical Name or Material Methylhydrazine sulfate
CAS 302-15-8
Molecular Formula CH3NHNH2·H2SO4
Formula Weight 144.15g/mol
MDL Number MFCD00035423
UN Number UN2811
Beilstein 3693575
Merck Index 14,6086
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1012206

 
alfa aesar™
L10122-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1012214

 
alfa aesar™
L10122-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 141° to 143°C
Quantity 5g
Chemical Name or Material Methylhydrazine sulfate
CAS 302-15-8
Assay 98%
Molecular Formula CH3NHNH2·H2SO4
Formula Weight 144.15g/mol
MDL Number MFCD00035423
UN Number UN2811
Beilstein 3693575
Merck Index 14,6086
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number MV7750000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.