Learn More
Methylgermanium trichloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04144909
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.
Solubility
Miscible with ether.
Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
Specifications
Methylgermanium trichloride | |
Colorless to Yellow | |
111°C | |
Chlorine-like | |
1.4685 | |
MFCD00013585 | |
UN2924 | |
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3 | |
Cl.Cl.Cl.C[GeH3] | |
200.06 | |
193.99 | |
Liquid |
993-10-2 | |
1.706 g/mL | |
10°C (50°F) | |
CH9Cl3Ge | |
CH3GeCl3 | |
10 g | |
Moisture sensitive | |
NNMJXXWNUALEKD-UHFFFAOYSA-N | |
trichloro(methyl)germane | |
70436 | |
97% |
Chemical Identifiers
993-10-2 | |
200.06 | |
NNMJXXWNUALEKD-UHFFFAOYSA-N | |
70436 | |
Cl.Cl.Cl.C[GeH3] |
CH9Cl3Ge | |
MFCD00013585 | |
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3 | |
trichloro(methyl)germane |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 213-604-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only