missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylgermanium trichloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04144902

 View more versions of this product

Catalog No. AA4144902



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.

Solubility
Miscible with ether.

Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
Specifications

Specifications

Methylgermanium trichloride
Colorless to Yellow
111°C
Chlorine-like
1.4685
MFCD00013585
UN2924
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
Cl.Cl.Cl.C[GeH3]
200.06
193.99
Liquid
993-10-2
1.706 g/mL
10°C (50°F)
CH9Cl3Ge
CH3GeCl3
0.5 g
Moisture sensitive
NNMJXXWNUALEKD-UHFFFAOYSA-N
trichloro(methyl)germane
70436
97%

Chemical Identifiers

993-10-2
200.06
NNMJXXWNUALEKD-UHFFFAOYSA-N
70436
Cl.Cl.Cl.C[GeH3]
CH9Cl3Ge
MFCD00013585
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
trichloro(methyl)germane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 213-604-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only