missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl tridecanoate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438200010
Specifications
Methyl tridecanoate | |
>110°C | |
Glass bottle | |
130°C to 132°C (4.0 Torr) | |
7°C | |
1731-88-0 | |
C14H28O2 | |
JNDDPBOKWCBQSM-UHFFFAOYSA-N | |
methyl tridecanoate | |
15608 | |
Liquid |
0.8600g/mL | |
Authentic | |
0.86 | |
Colorless | |
1g | |
96% min. (GC) | |
methyl n-tridecanoate, methyl tridecylate, methyltridecanoate, n-tridecanoic acid methyl ester, o2h463ring, tridecanoic acid methyl, tridecanoic acid methyl ester, tridecanoic acid, methyl ester, tridecanoic acid,methyl ester, unii-o2h463ring | |
CCCCCCCCCCCCC(=O)OC | |
228.38 | |
228.38 | |
97% |
Chemical Identifiers
1731-88-0 | |
228.38 | |
methyl n-tridecanoate, methyl tridecylate, methyltridecanoate, n-tridecanoic acid methyl ester, o2h463ring, tridecanoic acid methyl, tridecanoic acid methyl ester, tridecanoic acid, methyl ester, tridecanoic acid,methyl ester, unii-o2h463ring | |
methyl tridecanoate |
C14H28O2 | |
JNDDPBOKWCBQSM-UHFFFAOYSA-N | |
15608 | |
CCCCCCCCCCCCC(=O)OC |
Safety and Handling
EINECSNumber : 217-054-8