missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylethiocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164031000

 View more versions of this product

Catalog No. AC164031000


 

Specifications Description & Specifications

Specifications

Methylethiocyanate
100mL
1.068g/mL
Glass bottle
Liquid
99%
CH3SCN
MFCD00001830
15, 6200
Clear colorless to light yellow
556-64-9
Authentic
1.4670 to 1.4690 (20°C, 589nm)
≥98.5%
C2H3NS
73.11
03, 175

Chemical Identifiers

99%
CH3SCN
MFCD00001830
≥98.5%
C2H3NS
73.11
Methylethiocyanate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger