missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylethiocyanate, 99%, ACROS Organics™

$249.81 - $249.81

Chemical Identifiers

Assay Percent Range 99%
Molecular Formula C2H3NS
Linear Formula CH3SCN
Formula Weight 73.11
MDL Number MFCD00001830
Chemical Name or Material Methylethiocyanate
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC164031000 SDS Thermo Fisher Scientific
164031000
Glass bottle 100mL Each for $249.81
Specifications Description & Specifications

Chemical Identifiers

99%
CH3SCN
MFCD00001830
≥98.5%
C2H3NS
73.11
Methylethiocyanate

Specifications

Clear colorless to light yellow
556-64-9
Authentic
1.4670 to 1.4690 (20°C, 589nm)
99%
CH3SCN
MFCD00001830
15, 6200
≥98.5%
100mL
1.068g/mL
Glass bottle
Liquid
C2H3NS
73.11
03, 175
Methylethiocyanate