Please login to your online account to display your discounted pricing

Methylesuccinyl chloride, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC108400250

 View more versions of this product

Catalog No. AC108400250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 150.56
Quantity 25g
Beilstein 02, II, 553
Density 1.232g/mL
Packaging Glass bottle
Linear Formula CH3O2CCH2CH2COCl
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1490-25-1
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Chemical Name or Material Methylesuccinyl chloride
Molecular Formula C5H7ClO3
Assay Percent Range 97%
MDL Number MFCD00000750
Synonym Methyl 4-chloro-4-oxobutyrate

Specifications

Chemical Name or Material Methylesuccinyl chloride
Assay Percent Range 97%
Molecular Formula C5H7ClO3
Linear Formula CH3O2CCH2CH2COCl
Formula Weight 150.56
MDL Number MFCD00000750
Synonym Methyl 4-chloro-4-oxobutyrate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents