Please login to your online account to display your discounted pricing

Methylergometrine maleate, Tocris

Active metabolite of methysergide

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Methylergometrine maleate
Assay Percent Range >98%
Molecular Formula C20H25N3O2·C4H4O4
Formula Weight 455.51
Synonym [8β(S)]-9, 10-Didehydro-N-[1-(hydroxymethyl)propyl]-6-methylergoline-8-carboxamide maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

0549250

 
r & d systems
RDS0549250
50mg Each for $598.33

054950

 
r & d systems
RDS054950
50mg Each for $142.08
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 455.51
Assay Percent Range >98%
Molecular Formula C20H25N3O2·C4H4O4
Chemical Name or Material Methylergometrine maleate
CAS 57432-61-8
Synonym [8β(S)]-9, 10-Didehydro-N-[1-(hydroxymethyl)propyl]-6-methylergoline-8-carboxamide maleate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature