Please login to your online account to display your discounted pricing

Methylequinoxaline-6-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl quinoxaline-6-carboxylate
CAS 23088-23-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00102618
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDP00059EA

  SDS
fisher scientific company
RDP00059EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C10H8N2O2
MDL Number MFCD00102618
CAS 23088-23-5
Chemical Name or Material Methyl quinoxaline-6-carboxylate
Assay Percent Range 97%