Please login to your online account to display your discounted pricing

Methylequinoxaline-6-carboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RDP00059EA

 View more versions of this product

Catalog No. RDP00059EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C10H8N2O2
CAS 23088-23-5
MDL Number MFCD00102618
Assay Percent Range 97%
Chemical Name or Material Methyl quinoxaline-6-carboxylate

Specifications

Chemical Name or Material Methyl quinoxaline-6-carboxylate
CAS 23088-23-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00102618