missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylequinoline-6-carboxylate, 97%, Maybridge

$139.84 - $139.84

Chemical Identifiers

CAS 38896-30-9
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00956766
Chemical Name or Material Methyl quinoline-6-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
KM10245EB Fisher Scientific Company
KM10245EB
Amber Glass Bottle 25g Each for $139.84
Specifications Description & Specifications

Chemical Identifiers

38896-30-9
97%
187.2
Methyl quinoline-6-carboxylate
97.0
C11H9NO2
MFCD00956766
97%

Specifications

187.2
97.0
MFCD00956766
97%
Methyl quinoline-6-carboxylate
25g
38896-30-9
Amber Glass Bottle
C11H9NO2
97%