Please login to your online account to display your discounted pricing

Methylequinoline-6-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM10245DA

 View more versions of this product

Catalog No. KM10245DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.2
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C11H9NO2
MDL Number MFCD00956766
CAS 38896-30-9
Chemical Name or Material Methyl quinoline-6-carboxylate
Assay Percent Range 97%

Specifications

Chemical Name or Material Methyl quinoline-6-carboxylate
CAS 38896-30-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00956766