Learn More
Methyl propiolate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174040500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl propiolate | |
Colorless | |
98°C to 101°C | |
Authentic | |
Glass bottle | |
1.4070 to 1.409 | |
MFCD00008572 | |
02, I, 208 | |
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
methyl prop-2-ynoate | |
13536 | |
99% |
922-67-8 | |
0.9400g/mL | |
10°C | |
99% | |
C4H4O2 | |
HC∽CCO2CH3 | |
50 g | |
0.94 | |
Solubility in water: insoluble | |
COC(=O)C#C | |
84.07 | |
84.07 | |
Liquid |
Chemical Identifiers
922-67-8 | |
84.07 | |
IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
13536 | |
COC(=O)C#C |
C4H4O2 | |
MFCD00008572 | |
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
methyl prop-2-ynoate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio
GHS Signal Word: Danger
EINECSNumber : 213-083-5
RUO – Research Use Only